Assay Details
| Gene Symbol: CDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699055.3 | XP_005699112.1 | 5 - 6 | 630 | 90 | |
| XM_005699056.2 | XP_005699113.1 | 5 - 6 | 629 | 90 | ||
| XM_005699057.3 | XP_005699114.1 | 5 - 6 | 705 | 90 | ||
| XM_005699058.3 | XP_005699115.1 | 5 - 6 | 628 | 90 | ||
| GenBank mRNA | JI861808.1 | - | 4 - 5 | 491 | 90 | |
| JO595722.2 | - | 5 - 6 | 617 | 90 | ||
More Information
Set Membership: |
|

CDK1
Genomic Map