Assay Details
| Gene Symbol: PAX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052180.1 | XP_017907669.1 | 1 - 2 | 121 | 87 | |
| XM_018052181.1 | XP_017907670.1 | 1 - 2 | 43 | 87 | ||
| XM_018052183.1 | XP_017907672.1 | 1 - 2 | 43 | 87 | ||
More Information
Set Membership: |
|

PAX5
Genomic Map