Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966050.2 | XP_013821504.2 | 4 - 5 | 1125 | 87 | |
| XM_013966051.2 | XP_013821505.2 | 4 - 5 | 1125 | 87 | ||
| XM_013966054.2 | XP_013821508.2 | 4 - 5 | 1143 | 87 | ||
| XM_018052210.1 | XP_017907699.1 | 4 - 5 | 1125 | 87 | ||
| XM_018052211.1 | XP_017907700.1 | 2 - 3 | 761 | 87 | ||
More Information
Set Membership: |
|

TRMO
Genomic Map