Assay Details
| Gene Symbol: SPINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054432.1 | XP_017909921.1 | 10 - 11 | 1569 | 52 | |
| XM_018054433.1 | XP_017909922.1 | 10 - 11 | 1641 | 52 | ||
| XM_018054434.1 | XP_017909923.1 | 10 - 11 | 1668 | 52 | ||
| GenBank mRNA | KA326716.1 | - | 1 - 2 | 872 | 52 | |
More Information
Set Membership: |
|

SPINT1
Genomic Map