Assay Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056350.1 | XP_017911839.1 | 2 - 3 | 509 | 61 | |
| GenBank mRNA | JO438658.2 | - | 4 - 5 | 651 | 61 | |
| JO611101.2 | - | 2 - 3 | 485 | 61 | ||
More Information
Set Membership: |
|

ANKRD10
Genomic Map