Assay Details
| Gene Symbol: SPSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060389.1 | XP_017915878.1 | 2 - 3 | 1047 | 62 | |
| GenBank mRNA | JO619756.2 | - | 1 - 2 | 105 | 62 | |
More Information
Set Membership: |
|

SPSB1
Genomic Map