Assay Details
| Gene Symbol: LOC102182436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971736.2 | XP_013827190.2 | 4 - 5 | 884 | 87 | |
| XM_013971738.2 | XP_013827192.2 | 3 - 4 | 309 | 87 | ||
More Information
Set Membership: |
|

LOC102182436
Genomic Map