Assay Details
| Gene Symbol: LOC102176914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064681.1 | XP_017920170.1 | 1 - 2 | 147 | 81 | |
| GenBank mRNA | JO615614.2 | - | 6 - 7 | 1221 | 81 | |
More Information
Set Membership: |
|

LOC102176914
Genomic Map