Assay Details
| Gene Symbol: LOC102176914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064681.1 | XP_017920170.1 | 4 - 5 | 717 | 92 | |
| GenBank mRNA | JO615614.2 | - | 3 - 4 | 651 | 92 | |
More Information
Set Membership: |
|

LOC102176914
Genomic Map