Assay Details
| Gene Symbol: LOC102176914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064681.1 | XP_017920170.1 | 5 - 6 | 798 | 71 | |
| GenBank mRNA | JO615614.2 | - | 2 - 3 | 570 | 71 | |
More Information
Set Membership: |
|

LOC102176914
Genomic Map