Assay Details
| Gene Symbol: ARHGDIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065457.1 | XP_017920946.1 | 5 - 6 | 566 | 65 | |
| XM_018065458.1 | XP_017920947.1 | 5 - 6 | 896 | 65 | ||
| GenBank mRNA | JO498195.2 | - | 1 - 2 | 1500 | 65 | |
| JO632772.2 | - | 1 - 2 | 1556 | 65 | ||
More Information
Set Membership: |
|

ARHGDIA
Genomic Map