Assay Details
Gene Symbol: STAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064994.1 | XP_017920483.1 | 3 - 4 | 942 | 55 | |
XM_018064995.1 | XP_017920484.1 | 3 - 4 | 942 | 55 | ||
XM_018064996.1 | XP_017920485.1 | 3 - 4 | 870 | 55 | ||
XM_018064997.1 | XP_017920486.1 | 3 - 4 | 870 | 55 | ||
XM_018064998.1 | XP_017920487.1 | 3 - 4 | 942 | 55 | ||
XM_018064999.1 | XP_017920488.1 | 3 - 4 | 681 | 55 | ||
GenBank mRNA | JO623071.2 | - | 3 - 4 | 870 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
