Assay Details
| Gene Symbol: RPUSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041056.1 | XP_017896545.1 | 2 - 3 | 336 | 62 | |
| XM_018041057.1 | XP_017896546.1 | 1 - 2 | 683 | 62 | ||
| GenBank mRNA | JO432424.2 | - | 2 - 3 | 365 | 62 | |
More Information
Set Membership: |
|

RPUSD1
Genomic Map