Assay Details
| Gene Symbol: LOC102177399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041562.1 | XP_017897051.1 | 10 - 11 | 944 | 97 | |
| XM_018041563.1 | XP_017897052.1 | 10 - 11 | 861 | 97 | ||
| XM_018041564.1 | XP_017897053.1 | 9 - 10 | 874 | 97 | ||
| GenBank mRNA | JO616787.2 | - | 5 - 6 | 339 | 97 | |
More Information
Set Membership: |
|

LOC102177399
Genomic Map