Assay Details
| Gene Symbol: SIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042864.1 | XP_017898353.1 | 2 - 3 | 850 | 59 | |
| XM_018042865.1 | XP_017898354.1 | 2 - 3 | 824 | 59 | ||
| GenBank mRNA | JO580514.1 | - | 2 - 3 | 908 | 59 | |
More Information
Set Membership: |
|

SIPA1
Genomic Map