Assay Details
| Gene Symbol: ETS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052869.1 | XP_017908358.1 | 2 - 3 | 379 | 66 | |
| GenBank mRNA | JO427591.1 | - | 2 - 3 | 434 | 66 | |
| KA346233.1 | - | 8 - 9 | 3293 | 66 | ||
More Information
Set Membership: |
|

ETS2
Genomic Map