Assay Details
| Gene Symbol: DDX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676246.3 | XP_005676303.1 | 9 - 10 | 1733 | 67 | |
| GenBank mRNA | JO427178.1 | - | 5 - 6 | 1130 | 67 | |
More Information
Set Membership: |
|

DDX18
Genomic Map