Assay Details
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042069.1 | XP_017897558.1 | 2 - 3 | 494 | 89 | |
GenBank mRNA | KA346720.1 | - | 2 - 3 | 461 | 89 | |
KF515991.1 | - | 3 - 4 | 219 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
