Assay Details
| Gene Symbol: PPBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681750.3 | XP_005681807.2 | 1 - 2 | 155 | 77 | |
| GenBank mRNA | JO330674.2 | - | 1 - 2 | 226 | 77 | |
More Information
Set Membership: |
|

PPBP
Genomic Map