Assay Details
| Gene Symbol: SMAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682962.2 | XP_005683019.1 | 5 - 6 | 1316 | 74 | |
| XM_018050347.1 | XP_017905836.1 | 5 - 6 | 1184 | 74 | ||
More Information
Set Membership: |
|

SMAD5
Genomic Map