Assay Details
| Gene Symbol: TRAF3IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684565.3 | XP_005684622.2 | 8 - 9 | 2026 | 70 | |
| XM_013966430.2 | XP_013821884.1 | 3 - 4 | 256 | 70 | ||
| XM_018053057.1 | XP_017908546.1 | 7 - 8 | 1584 | 70 | ||
| XM_018053059.1 | XP_017908548.1 | 3 - 4 | 411 | 70 | ||
| GenBank mRNA | KA321345.1 | - | 8 - 9 | 1845 | 70 | |
More Information
Set Membership: |
|

TRAF3IP2
Genomic Map