Assay Details
| Gene Symbol: AQP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685715.3 | XP_005685772.1 | 4 - 5 | 757 | 73 | |
| GenBank mRNA | JO420295.2 | - | 2 - 3 | 1377 | 73 | |
| JO563342.2 | - | 1 - 2 | 116 | 73 | ||
More Information
Set Membership: |
|

AQP9
Genomic Map