Assay Details
| Gene Symbol: POP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058275.1 | XP_017913764.1 | 13 - 14 | 1985 | 63 | |
| GenBank mRNA | JT683217.1 | - | 7 - 8 | 1287 | 63 | |
More Information
Set Membership: |
|

POP1
Genomic Map