Assay Details
| Gene Symbol: HMBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689536.3 | XP_005689593.1 | 5 - 6 | 455 | 62 | |
| GenBank mRNA | JO595643.2 | - | 5 - 6 | 463 | 62 | |
| JT708025.1 | - | 7 - 8 | 623 | 62 | ||
More Information
Set Membership: |
|

HMBS
Genomic Map