Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062379.1 | XP_017917868.1 | 12 - 13 | 1672 | 56 | |
| XM_018062380.1 | XP_017917869.1 | 12 - 13 | 1469 | 56 | ||
| XM_018062381.1 | XP_017917870.1 | 12 - 13 | 1667 | 56 | ||
| GenBank mRNA | KA340977.1 | - | 12 - 13 | 1435 | 56 | |
More Information
Set Membership: |
|

ACP7
Genomic Map