Assay Details
| Gene Symbol: ATP5MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693700.3 | XP_005693757.1 | 3 - 4 | 274 | 98 | |
| XM_013972310.2 | XP_013827764.1 | 3 - 4 | 266 | 98 | ||
More Information
Set Membership: |
|

ATP5MC1
Genomic Map