Assay Details
| Gene Symbol: ADAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065957.1 | XP_017921446.1 | 9 - 10 | 1075 | 75 | |
| XM_018065958.1 | XP_017921447.1 | 9 - 10 | 1000 | 75 | ||
| XM_018065959.1 | XP_017921448.1 | 8 - 9 | 842 | 75 | ||
| XM_018065960.1 | XP_017921449.1 | 9 - 10 | 1075 | 75 | ||
| XM_018065961.1 | XP_017921450.1 | 9 - 10 | 1075 | 75 | ||
| XM_018065962.1 | XP_017921451.1 | 9 - 10 | 1075 | 75 | ||
More Information
Set Membership: |
|

ADAL
Genomic Map