Assay Details
| Gene Symbol: HYAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695926.2 | XP_005695983.1 | 2 - 3 | 1194 | 66 | |
| XM_018038368.1 | XP_017893857.1 | 2 - 3 | 1324 | 66 | ||
| GenBank mRNA | JO578408.1 | - | 2 - 3 | 879 | 66 | |
| JO635170.2 | - | 2 - 3 | 732 | 66 | ||
More Information
Set Membership: |
|

HYAL2
Genomic Map