Assay Details
| Gene Symbol: DALRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066748.1 | XP_017922237.1 | 6 - 7 | 998 | 67 | |
| XM_018066749.1 | XP_017922238.1 | 5 - 6 | 937 | 67 | ||
| XM_018066750.1 | XP_017922239.1 | 6 - 7 | 998 | 67 | ||
More Information
Set Membership: |
|

DALRD3
Genomic Map