Assay Details
| Gene Symbol: C23H6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696265.3 | XP_005696322.2 | 7 - 8 | 1070 | 100 | |
| XM_005696266.3 | XP_005696323.2 | 6 - 7 | 997 | 100 | ||
| XM_013973851.2 | XP_013829305.2 | 7 - 8 | 1298 | 100 | ||
| XM_013973852.2 | XP_013829306.2 | 5 - 6 | 690 | 100 | ||
| GenBank mRNA | JO424772.1 | - | 7 - 8 | 1108 | 100 | |
| KA336915.1 | - | 2 - 3 | 4180 | 100 | ||
| Gene Symbol: CRYZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO501164.2 | - | 1341 | 100 | ||
More Information
Set Membership: |
|

C23H6orf89
Genomic Map