Assay Details
| Gene Symbol: LOC108634897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045228.1 | XP_017900717.1 | 249 | 69 | ||
| GenBank mRNA | JO436090.2 | - | 249 | 69 | ||
| Gene Symbol: FAM120C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043858.1 | XP_017899347.1 | 10 - 11 | 2417 | 69 | |
More Information
Set Membership: |
|

LOC108634897
Genomic Map