Assay Details
Gene Symbol: LOC108634897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045228.1 | XP_017900717.1 | 362 | 101 | ||
GenBank mRNA | JO436090.2 | - | 362 | 101 |
Gene Symbol: FAM120C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043858.1 | XP_017899347.1 | 11 - 12 | 2530 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
