Assay Details
Gene Symbol: LOC108634897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045228.1 | XP_017900717.1 | 1 | 768 | 97 | |
GenBank mRNA | JO436090.2 | - | 768 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
