Assay Details
| Gene Symbol: LOC108634897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045228.1 | XP_017900717.1 | 1 - 2 | 907 | 83 | |
| GenBank mRNA | JO436090.2 | - | 907 | 83 | ||
| JO509305.2 | - | 1 - 2 | 179 | 83 | ||
More Information
Set Membership: |
|

LOC108634897
Genomic Map