Assay Details
Gene Symbol: LOC108634897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045228.1 | XP_017900717.1 | 2 - 3 | 958 | 74 | |
GenBank mRNA | JO436090.2 | - | 958 | 74 | ||
JO509305.2 | - | 2 - 3 | 230 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
