Assay Details
| Gene Symbol: STK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709527.3 | XP_005709584.2 | 5 - 6 | 888 | 61 | |
| GenBank mRNA | KA323801.1 | - | 3 - 4 | 682 | 61 | |
More Information
Set Membership: |
|

STK19
Genomic Map