Assay Details
| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039978.1 | XP_017895467.1 | 6 - 7 | 945 | 63 | |
| GenBank mRNA | JO420506.2 | - | 6 - 7 | 985 | 63 | |
More Information
Set Membership: |
|

AGXT
Genomic Map