Assay Details
| Gene Symbol: DUSP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048465.1 | XP_017903954.1 | 6 - 7 | 1607 | 69 | |
| XM_018048466.1 | XP_017903955.1 | 6 - 7 | 1277 | 69 | ||
| XM_018048467.1 | XP_017903956.1 | 6 - 7 | 1265 | 69 | ||
| XM_018048468.1 | XP_017903957.1 | 6 - 7 | 1229 | 69 | ||
More Information
Set Membership: |
|

DUSP16
Genomic Map