Assay Details
| Gene Symbol: RASSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057361.1 | XP_017912850.1 | 7 - 8 | 718 | 67 | |
| XM_018057362.1 | XP_017912851.1 | 7 - 8 | 718 | 67 | ||
| GenBank mRNA | JO340661.2 | - | 3 - 4 | 1082 | 67 | |
| JO610706.2 | - | 3 - 4 | 897 | 67 | ||
More Information
Set Membership: |
|

RASSF2
Genomic Map