Assay Details
| Gene Symbol: UBE2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057933.1 | XP_017913422.1 | 5 - 6 | 578 | 64 | |
| GenBank mRNA | KA353397.1 | - | 5 - 6 | 612 | 64 | |
More Information
Set Membership: |
|

UBE2C
Genomic Map