Assay Details
| Gene Symbol: LOC102178921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968985.2 | XP_013824439.1 | 10 - 11 | 1885 | 69 | |
| GenBank mRNA | JO439710.2 | - | 3 - 4 | 552 | 69 | |
More Information
Set Membership: |
|

LOC102178921
Genomic Map