Assay Details
| Gene Symbol: LOC102178921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968985.2 | XP_013824439.1 | 6 - 7 | 1295 | 72 | |
| XM_018058054.1 | XP_017913543.1 | 6 - 7 | 1299 | 72 | ||
More Information
Set Membership: |
|

LOC102178921
Genomic Map