Assay Details
| Gene Symbol: LOC102178921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968985.2 | XP_013824439.1 | 7 - 8 | 1445 | 101 | |
| XM_018058054.1 | XP_017913543.1 | 7 - 8 | 1449 | 101 | ||
| GenBank mRNA | JO439710.2 | - | 6 - 7 | 992 | 101 | |
More Information
Set Membership: |
|

LOC102178921
Genomic Map