Assay Details
| Gene Symbol: LOC102178921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968985.2 | XP_013824439.1 | 9 - 10 | 1687 | 103 | |
| XM_018058054.1 | XP_017913543.1 | 9 - 10 | 1691 | 103 | ||
| GenBank mRNA | JO439710.2 | - | 4 - 5 | 750 | 103 | |
More Information
Set Membership: |
|

LOC102178921
Genomic Map