Assay Details
| Gene Symbol: C1H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969425.2 | XP_013824879.1 | 2 - 3 | 661 | 92 | |
| GenBank mRNA | JO319577.2 | - | 2 - 3 | 434 | 92 | |
| JO404811.2 | - | 1 - 2 | 437 | 92 | ||
| JO404812.2 | - | 1 - 2 | 437 | 92 | ||
| JO435055.2 | - | 1 - 2 | 1029 | 92 | ||
| JO513082.2 | - | 2 - 3 | 472 | 92 | ||
| JO565483.2 | - | 2 - 3 | 380 | 92 | ||
| JO568505.2 | - | 2 - 3 | 380 | 92 | ||
| JO585668.2 | - | 1 - 2 | 1009 | 92 | ||
| JT716482.1 | - | 3 - 4 | 682 | 92 | ||
More Information
Set Membership: |
|

C1H3orf38
Genomic Map