Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059286.1 | XP_017914775.1 | 9 - 10 | 1619 | 57 | |
| XM_018059287.1 | XP_017914776.1 | 8 - 9 | 1437 | 57 | ||
| XM_018059288.1 | XP_017914777.1 | 9 - 10 | 1686 | 57 | ||
More Information
Set Membership: |
|

WT1
Genomic Map