Assay Details
| Gene Symbol: SLC7A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062274.1 | XP_017917763.1 | 6 - 7 | 1002 | 57 | |
| XM_018062275.1 | XP_017917764.1 | 6 - 7 | 1051 | 57 | ||
| XM_018062276.1 | XP_017917765.1 | 6 - 7 | 1002 | 57 | ||
| XM_018062277.1 | XP_017917766.1 | 6 - 7 | 969 | 57 | ||
| GenBank mRNA | JO623261.2 | - | 6 - 7 | 1000 | 57 | |
| JO623263.2 | - | 6 - 7 | 1000 | 57 | ||
More Information
Set Membership: |
|

SLC7A10
Genomic Map