Assay Details
| Gene Symbol: LOC108633216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045500.1 | XP_017900989.1 | 10 - 11 | 1580 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC108633216| Gene Symbol: LOC108633216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045500.1 | XP_017900989.1 | 10 - 11 | 1580 | 92 | |
Set Membership: |
|