Assay Details
Gene Symbol: LOC108635343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045525.1 | XP_017901014.1 | 249 | 85 |
Gene Symbol: LOC108635346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045526.1 | XP_017901015.1 | 2 - 3 | 176 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
