Assay Details
| Gene Symbol: PLA2G5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067054.1 | XP_017922543.1 | 4 - 5 | 334 | 89 | |
| GenBank mRNA | JO625498.2 | - | 1 - 2 | 168 | 89 | |
More Information
Set Membership: |
|

PLA2G5
Genomic Map